Learn More
p-Phenylenediamine, 97%
CAS: 106-50-3 | C6H8N2 | 108.144 g/mol
Supplier: Thermo Scientific Chemicals A156800I
Description
p-Phenylenediamine is used as a precursor to aramid plastics, Kevlar fibers and to urethane polymers. It serves as a developing agent in photography, antioxidants in rubber products and as a vulcanization accelerator. It is used as a henna surrogate for temporary tattoos and as a histological stain for lipids such as myelin. It is an anti-fade reagent used in fluorescence microscopy to retard photobleaching of fluorescein and other fluorescent dyes. Furthermore, it is also an ingredient in hair dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsp-Phenylenediamine is used as a precursor to aramid plastics, Kevlar fibers and to urethane polymers. It serves as a developing agent in photography, antioxidants in rubber products and as a vulcanization accelerator. It is used as a henna surrogate for temporary tattoos and as a histological stain for lipids such as myelin. It is an anti-fade reagent used in fluorescence microscopy to retard photobleaching of fluorescein and other fluorescent dyes. Furthermore, it is also an ingredient in hair dyes.
Solubility
Soluble in alcohol, chloroform, ether and hot benzene.
Notes
Store in cool place. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
Specifications
| p-Phenylenediamine | |
| 138°C to 142°C | |
| 156°C (312°F) | |
| Weak | |
| MFCD00007901 | |
| UN1673 | |
| 14,7285 | |
| Soluble in alcohol,chloroform,ether and hot benzene. | |
| C1=CC(=CC=C1N)N | |
| 108.144 | |
| CHEBI:51403 | |
| 97% |
| 106-50-3 | |
| 267°C to 271°C | |
| 97% | |
| C6H8N2 | |
| 5000 g | |
| 742029 | |
| p-phenylenediamine, 1,4-benzenediamine, 1,4-diaminobenzene, 1,4-phenylenediamine, 4-aminoaniline, para-phenylenediamine, p-aminoaniline, p-diaminobenzene, orsin, 4-phenylenediamine | |
| CBCKQZAAMUWICA-UHFFFAOYSA-N | |
| benzene-1,4-diamine | |
| 7814 | |
| 108.14 |
Chemical Identifiers
| 106-50-3 | |
| 108.144 | |
| CBCKQZAAMUWICA-UHFFFAOYSA-N | |
| 7814 | |
| benzene-1,4-diamine |
| C6H8N2 | |
| MFCD00007901 | |
| p-phenylenediamine, 1,4-benzenediamine, 1,4-diaminobenzene, 1,4-phenylenediamine, 4-aminoaniline, para-phenylenediamine, p-aminoaniline, p-diaminobenzene, orsin, 4-phenylenediamine | |
| CHEBI:51403 | |
| C1=CC(=CC=C1N)N |
Safety and Handling
GHS H Statement
H301-H311-H331-H319-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye irritation.
May cause an allergic skin reaction.
P261-P264b-P270-P271-P272-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P333+P313-P361-P363-P501c
H301+H311+H331-H317-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: PHENYLENEDIAMINES
EINECSNumber : 203-404-7
RTECSNumber : SS8050000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only