missing translation for 'onlineSavingsMsg'
Learn More

p-Phenylenediamine, 97%

CAS: 106-50-3 | C6H8N2 | 108.144 g/mol

Supplier:  Thermo Scientific Chemicals A156800B

 View more versions of this product

Catalog No. AAA156800B


Only null left
Explore available promotions

Description

Description

p-Phenylenediamine is used as a precursor to aramid plastics, Kevlar fibers and to urethane polymers. It serves as a developing agent in photography, antioxidants in rubber products and as a vulcanization accelerator. It is used as a henna surrogate for temporary tattoos and as a histological stain for lipids such as myelin. It is an anti-fade reagent used in fluorescence microscopy to retard photobleaching of fluorescein and other fluorescent dyes. Furthermore, it is also an ingredient in hair dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Phenylenediamine is used as a precursor to aramid plastics, Kevlar fibers and to urethane polymers. It serves as a developing agent in photography, antioxidants in rubber products and as a vulcanization accelerator. It is used as a henna surrogate for temporary tattoos and as a histological stain for lipids such as myelin. It is an anti-fade reagent used in fluorescence microscopy to retard photobleaching of fluorescein and other fluorescent dyes. Furthermore, it is also an ingredient in hair dyes.

Solubility
Soluble in alcohol, chloroform, ether and hot benzene.

Notes
Store in cool place. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
Specifications

Specifications

p-Phenylenediamine
138°C to 142°C
156°C (312°F)
Weak
MFCD00007901
UN1673
14,7285
Soluble in alcohol,chloroform,ether and hot benzene.
C1=CC(=CC=C1N)N
108.144
CHEBI:51403
97%
106-50-3
267°C to 271°C
97%
C6H8N2
1000 g
742029
p-phenylenediamine, 1,4-benzenediamine, 1,4-diaminobenzene, 1,4-phenylenediamine, 4-aminoaniline, para-phenylenediamine, p-aminoaniline, p-diaminobenzene, orsin, 4-phenylenediamine
CBCKQZAAMUWICA-UHFFFAOYSA-N
benzene-1,4-diamine
7814
108.14

Chemical Identifiers

106-50-3
108.144
CBCKQZAAMUWICA-UHFFFAOYSA-N
7814
benzene-1,4-diamine
C6H8N2
MFCD00007901
p-phenylenediamine, 1,4-benzenediamine, 1,4-diaminobenzene, 1,4-phenylenediamine, 4-aminoaniline, para-phenylenediamine, p-aminoaniline, p-diaminobenzene, orsin, 4-phenylenediamine
CHEBI:51403
C1=CC(=CC=C1N)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H319-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye irritation.
May cause an allergic skin reaction.

P261-P264b-P270-P271-P272-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P333+P313-P361-P363-P501c

H301+H311+H331-H317-H319

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: PHENYLENEDIAMINES

EINECSNumber : 203-404-7

RTECSNumber : SS8050000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only