missing translation for 'onlineSavingsMsg'
Learn More

p-Nitrophenyl phosphate disodium salt hexahydrate, >99%, MP Biomedicals™

Catalog No. ICN10087801
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10087801 1 g
ICN10087805 5 g
ICN10087850 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10087801 Supplier MP Biomedicals Inc Supplier No. 0210087801

Chemical Identifiers

CAS 4264-83-9
Molecular Formula C6H4NNa2O6P
Molecular Weight (g/mol) 263.05
MDL Number MFCD00007319
InChI Key VIYFPAMJCJLZKD-UHFFFAOYSA-L
Synonym pnpp, disodium 4-nitrophenylphosphate, sodium 4-nitrophenyl phosphate, disodium 4-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, disodium salt, disodium p-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2, p-nitrophenyl phosphate, pnpp liquid substrate system
PubChem CID 77949
SMILES [Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1

Specifications

Color White to Yellow
Assay Percent Range >99%
Quantity 1 g
IUPAC Name disodium;(4-nitrophenyl) phosphate
Formula Weight 371
Physical Form Crystalline Powder
Chemical Name or Material p-Nitrophenyl phosphate disodium salt hexahydrate