missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Naphtholbenzein Indicator, 1% in Titration Solvent, Reagents
Specifications
Specifications
| Chemical Name or Material | p-Naphtholbenzein Indicator, 1% in Titration Solvent ASTM Methods D974, D3242, D3339 |
| Boiling Point | 90°C |
| CAS | 108-88-3, 67-63-0, 7732-18-5 |
| Packaging | Glass Bottle |
| Specific Gravity | 0.8245 |
| Color | Dark Red |
| Synonym | alpha-Naphtholbenzein, para-Naphtholbenzein, 1-Naphtholbenzein |
| Quantity | 120 mL |
| Physical Form | Liquid |
| Grade | ASTM |