missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
p-Dioxane, Anhydrous, 99.9% min., DriSolv, MilliporeSigma™
Supplier: Technidata DX20927
Specifications
p-Dioxane | |
123-91-1 | |
1.03g/cm3 (20°C) | |
1.7 to 25.2%(V) | |
Liquid | |
C4H8O2 | |
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin | |
C1COCCO1 | |
88.106 | |
CHEBI:47032 |
SeccoSolv™, Dried | |
Colorless | |
100 mL | |
300°C | |
99.9% min. | |
41 hPa (20°C) | |
RYHBNJHYFVUHQT-UHFFFAOYSA-N | |
1,4-dioxane | |
31275 | |
88.11 |
Chemical Identifiers
123-91-1 | |
88.106 | |
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin | |
CHEBI:47032 | |
C1COCCO1 |
C4H8O2 | |
RYHBNJHYFVUHQT-UHFFFAOYSA-N | |
31275 | |
1,4-dioxane |
Safety and Handling
Recommended Storage : Store at +15°C to +25°C