missing translation for 'onlineSavingsMsg'
Learn More

p-Dioxane, 99.9% min., OmniSolv, MilliporeSigma™

Catalog No. MDX20916
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MDX20916 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MDX20916 Supplier Technidata Supplier No. DX20916

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 123-91-1
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
Synonym dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
PubChem CID 31275
ChEBI CHEBI:47032
IUPAC Name 1,4-dioxane
SMILES C1COCCO1

Specifications

Quantity 1 L
Assay Percent Range 99.0%, min.
Linear Formula -O(CH2)2O(CH2)2-
Formula Weight 88.1
Grade ACS/NF/USP
Chemical Name or Material p-Dioxane, OmniSolv™