missing translation for 'onlineSavingsMsg'
Learn More

p-Dioxane, 99.0% min., ACS, MilliporeSigma™

Catalog No. MDX20906
Change view
Click to view available options
Quantity:
500 mL
4 L
207 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MDX20906 500 mL
MDX20903 4 L
MDX209030 207 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MDX20906 Supplier Technidata Supplier No. DX20906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 123-91-1
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
Synonym dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
PubChem CID 31275
ChEBI CHEBI:47032
IUPAC Name 1,4-dioxane
SMILES C1COCCO1

Specifications

Quantity 500 mL
Assay Percent Range 99.0%, min.
Linear Formula -O(CH2)2O(CH2)2-
Formula Weight 88.1
Grade ACS/NF/USP
Chemical Name or Material p-Dioxane, For ACS analysis