missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich p-Cymene

Catalog No. C1214524L
Encompass
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1214524L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C1214524L Supplier Sigma Aldrich Supplier No. C1214524L
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

p-Cymene is an alkyl aromatic hydrocarbon mainly used to study oxidation reactions under various conditions to synthesize highly valuable intermediates and building-blocks such as cymene hydroperoxide, 4-methyl acetophenone, 4-isopropyl benzyl alcohol, 4-isopropyl benzaldehyde and 4-isopropyl benzoic acid etc.

Specifications

CAS 99-87-6
Density 0.86 g/mL (at 25°C (literature))
Boiling Point 176°C to 178°C (lit.)
Molecular Formula C10H14
Refractive Index n20/D 1.490 (literature)
Linear Formula CH3C6H4CH(CH3)2
MDL Number MFCD00008893
Quantity 4 L
Synonym 1-Isopropyl-4-methylbenzene; 4-Isopropyltoluene
Molecular Weight (g/mol) 134.22
Percent Purity 99%
Show More Show Less