missing translation for 'onlineSavingsMsg'
Learn More

p-Chloranil, 97%

Catalog No. AAA1349514
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1349514 25 g
AAA1349522 100 g
AAA1349536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1349514 Supplier Thermo Scientific Chemicals Supplier No. A1349514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 118-75-2 | C6Cl4O2 | 245.86 g/mol

p-Chloranil is used as a dye intermediate, oxidizing agent, vulcanizing agent and dehydrogenation reagent. It is also used to make chloranil electrodes for pH measurements. Further, it serves as a hydrogen acceptor and used for the aromatization reactions such as conversion of cyclohexadienes to the benzene derivatives. It is used to test the secondary amine. In addition to this, it is employed as a precursor to prepare diaziquone, which is used as a cancer chemotherapeutic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Chloranil is used as a dye intermediate, oxidizing agent, vulcanizing agent and dehydrogenation reagent. It is also used to make chloranil electrodes for pH measurements. Further, it serves as a hydrogen acceptor and used for the aromatization reactions such as conversion of cyclohexadienes to the benzene derivatives. It is used to test the secondary amine. In addition to this, it is employed as a precursor to prepare diaziquone, which is used as a cancer chemotherapeutic agent.

Solubility
Soluble in ether and acetone. Slightly soluble in chloroform, dimethylformamide, solvent naphtha, benzene, carbon disulfide and carbon tetrachloride. Insoluble in water, cold petroleum ether, methanol, dibutyl phthalate and cold alcohol.

Notes
Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 118-75-2
Molecular Formula C6Cl4O2
Molecular Weight (g/mol) 245.86
MDL Number MFCD00001594
InChI Key UGNWTBMOAKPKBL-UHFFFAOYSA-N
Synonym chloranil, p-chloranil, tetrachloro-p-benzoquinone, tetrachloro-1,4-benzoquinone, tetrachlorobenzoquinone, spergon, coversan, reranil, vulklor, 2,3,5,6-tetrachloro-1,4-benzoquinone
PubChem CID 8371
ChEBI CHEBI:36703
IUPAC Name 2,3,5,6-tetrachlorocyclohexa-2,5-diene-1,4-dione
SMILES ClC1=C(Cl)C(=O)C(Cl)=C(Cl)C1=O

Specifications

Melting Point ∼290°C (sublimation)
Density 1.97
Quantity 25 g
UN Number UN3077
Beilstein 393006
Merck Index 14,2078
Solubility Information Soluble in ether and acetone. Slightly soluble in chloroform,dimethylformamide,solvent naphtha,benzene,carbon disulfide and carbon tetrachloride. Insoluble in water,cold petroleum ether,methanol,dibutyl phthalate and cold alcohol.
Formula Weight 245.88
Percent Purity 97%
Chemical Name or Material p-Chloranil
Hazard Category H315-H319-H331
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P311-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 204-274-4
RTECSNumber DK6825000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only