missing translation for 'onlineSavingsMsg'
Learn More

p-Acetoacetanisidide 98.0+%, TCI America™

Catalog No. p-7117364
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A130725G 25 g
A1307500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A130725G Supplier TCI America Supplier No. A130725G
Only null left

Chemical Identifiers

CAS 5437-98-9
Molecular Formula C11H13NO3
Molecular Weight (g/mol) 207.229
MDL Number MFCD00008783
InChI Key SWAJJKROCOJICG-UHFFFAOYSA-N
Synonym n-4-methoxyphenyl-3-oxobutanamide, p-acetoacetanisidide, 4'-methoxyacetoacetanilide, acetoacet-p-anisidide, p-acetoacetanisidine, para-acetoacetanisidide, butanamide, n-4-methoxyphenyl-3-oxo, n-4-methoxyphenyl-3-oxo-butyramide, p-acetoacetaniside, n-4-methoxyphenyl acetoacetamide
PubChem CID 21576
IUPAC Name N-(4-methoxyphenyl)-3-oxobutanamide
SMILES CC(=O)CC(=O)NC1=CC=C(C=C1)OC

Specifications

Melting Point 117°C
Color Blue-White
Quantity 25 g
Formula Weight 207.23
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material p-Acetoacetanisidide
EINECSNumber (3)-0689
RTECSNumber AK4598010
TSCA Yes