missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ozagrel Hydrochloride Hydrate 98.0+%, TCI America™
Supplier: TCI America O04195G
Specifications
| Ozagrel Hydrochloride Hydrate | |
| 217°C | |
| C13H13ClN2O2 | |
| 5 g | |
| CWKFWBJJNNPGAM-IPZCTEOASA-N | |
| (E)-3-[4-(imidazol-1-ylmethyl)phenyl]prop-2-enoic acid;hydrochloride | |
| 6438130 | |
| 264.71 | |
| Crystalline Powder |
| 78712-43-3 | |
| White | |
| MFCD06795644 | |
| (E)-3-[4-(Imidazol-1-ylmethyl)phenyl]propenoic Acid Hydrochloride, trans-4-[(1-Imidazolyl)methyl]cinnamic Acid Hydrochloride | |
| C1=CC(=CC=C1CN2C=CN=C2)C=CC(=O)O.Cl | |
| 264.709 | |
| CHEBI:31954 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 78712-43-3 | |
| 264.709 | |
| CWKFWBJJNNPGAM-IPZCTEOASA-N | |
| 6438130 | |
| (E)-3-[4-(imidazol-1-ylmethyl)phenyl]prop-2-enoic acid;hydrochloride |
| C13H13ClN2O2 | |
| MFCD06795644 | |
| (E)-3-[4-(Imidazol-1-ylmethyl)phenyl]propenoic Acid Hydrochloride, trans-4-[(1-Imidazolyl)methyl]cinnamic Acid Hydrochloride | |
| CHEBI:31954 | |
| C1=CC(=CC=C1CN2C=CN=C2)C=CC(=O)O.Cl |
Safety and Handling
RTECSNumber : UD3380000
TSCA : No