missing translation for 'onlineSavingsMsg'
Learn More

Ozagrel Hydrochloride Hydrate 98.0+%, TCI America™

Catalog No. O04191G
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
O04191G 1 g
O04195G 5 g
2 options

Catalog No. O04191G

Supplier: TCI America O04191G

Only null left

Chemical Identifiers

CAS 78712-43-3
Molecular Formula C13H13ClN2O2
Molecular Weight (g/mol) 264.709
MDL Number MFCD06795644
InChI Key CWKFWBJJNNPGAM-IPZCTEOASA-N
Synonym (E)-3-[4-(Imidazol-1-ylmethyl)phenyl]propenoic Acid Hydrochloride, trans-4-[(1-Imidazolyl)methyl]cinnamic Acid Hydrochloride
PubChem CID 6438130
ChEBI CHEBI:31954
IUPAC Name (E)-3-[4-(imidazol-1-ylmethyl)phenyl]prop-2-enoic acid;hydrochloride
SMILES C1=CC(=CC=C1CN2C=CN=C2)C=CC(=O)O.Cl

Specifications

Melting Point 217°C
Color White
Quantity 1 g
Formula Weight 264.71
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ozagrel Hydrochloride Hydrate
RTECSNumber UD3380000
TSCA No