missing translation for 'onlineSavingsMsg'
Learn More

Oxytocin, 96%

Catalog No. p-7042699
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63421LB0 5 mg
AAJ63421MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63421LB0 Supplier Thermo Scientific Chemicals Supplier No. J63421LB0

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 50-56-6 | C45H67F3N12O14S2 | 1121.216 g/mol

Oxytocin is used as a peptide hormone that acts predominantly as a neurotransmitter; stimulates uterine contraction and lactation, increases Na+ excretion; stimulates myometrial GTPase and phospholipase C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oxytocin is used as a peptide hormone that acts predominantly as a neurotransmitter; stimulates uterine contraction and lactation, increases Na+ excretion; stimulates myometrial GTPase and phospholipase C.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 50-56-6
Molecular Formula C45H67F3N12O14S2
Molecular Weight (g/mol) 1121.216
MDL Number MFCD00076731
InChI Key HCWNGYCIMQIGKC-BXUJZNQYSA-N
Synonym oxytocin acetate syntocinon /
PubChem CID 87795236
IUPAC Name (2S)-1-[(4R,7S,10S,13S,16S,19R)-19-amino-7-(2-amino-2-oxoethyl)-10-(3-amino-3-oxopropyl)-13-[(2S)-butan-2-yl]-16-[(4-hydroxyphenyl)methyl]-6,9,12,15,18-pentaoxo-1,2-dithia-5,8,11,14,17-pentazacycloicosane-4-carbonyl]-N-[(2S)-1-[(2-amino-2-oxoethyl)amino]-
SMILES CCC(C)C1C(=O)NC(C(=O)NC(C(=O)NC(CSSCC(C(=O)NC(C(=O)N1)CC2=CC=C(C=C2)O)N)C(=O)N3CCCC3C(=O)NC(CC(C)C)C(=O)NCC(=O)N)CC(=O)N)CCC(=O)N.C(=O)(C(F)(F)F)O

Specifications

Color White
Quantity 5 mg
UN Number UN2811
Merck Index 14,6979
Solubility Information Soluble in water.
Formula Weight 1007.19
Percent Purity 96%
Physical Form Lyophilized Powder
Chemical Name or Material Oxytocin
Hazard Category H300-H361
Hazard Statement GHS H Statement
H300-H361
Fatal if swallowed.
Suspected of damaging fertility or the unborn child.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P310-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 200-048-4
RTECSNumber RS7534000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only