missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Oxytetracycline Dihydrate 95%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Type | Oxytetracycline Dihydrate |
| Physical Form | Crystalline Powder |
| Infrared Spectrum | Authentic |
| Packaging | Glass Bottle |
| Specific Rotation | On anhydrous substance: -203° to -216° (20°C, 589nm) (c=1, 0.1M HCl) |
| Water | 4 to 8% |
| Color | Yellow |
| Quantity | 100 g |
| Assay Percent Range | 95% |
| Merck Index | 14, 6976 |
| Show More |
Safety and Handling
| Hazard Category | LIGHT SENSITIVE |
| Hazard Statement | GHS H Statement: Causes serious eye irritation. Suspected of damaging the unborn child. |
| Precautionary Statement | GHS P Statement: Use personal protective equipment as required. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
RUO – Research Use Only