missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ OxyBlot™ Protein Standard

Catalog No. S7151
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S7151 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S7151 Supplier MilliporeSigma™ Supplier No. S7151

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Each vial contains 150μL of OxyBlot Protein Standard, which is sufficient for 60 minigels

Internal control for the electrophoresis, western transfer and immunodetection steps of the OxyBlot procedure

  • Composed of five proteins (phosphorylase B, BSA, ovalbumin, carbonic anhydrase, trypsin inhibitor) containing 1-3 dinitrophenylhydrazone (DNP) residues per protein
  • These attached DNP moieties permit the visualization of the proteins by the detection system of the OxyBlot Protein Oxidation Detection Kit

Specifications

Content And Storage Prior to first use, store at 4°C. Store the aliquots at -15°C to -25°C
For Use With (Application) Western Blotting
Quantity 150 μL