missing translation for 'onlineSavingsMsg'
Learn More

Oxonol Yellow K, TCI America™

Catalog No. o028425g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O028425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O028425G Supplier TCI America Supplier No. O028425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 82407-55-4
Molecular Formula C21H18N4O8S2
MDL Number MFCD02093486
Synonym 4,4′C-Methylidynebis[1-(4-sulfophenyl)-3-methyl-5-pyrazolone] Dipotassium Salt

Specifications

Color Yellow
Quantity 25 g
Formula Weight 594.70
Physical Form Crystalline Powder
Chemical Name or Material Oxonol Yellow K