missing translation for 'onlineSavingsMsg'
Learn More

Oxone(R), Monopersulfate Compound, Spectrum™ Chemical

Catalog No. 18604919
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
25 kg
Packaging:
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604919 100 g Poly Bottle
18604920 1 kg Poly Bottle
18604921 5 kg Poly Pail
18604922 25 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18604919 Supplier Spectrum Chemical Mfg Cor Supplier No. O1306100GM

O1306, 37222-66-5, 2KHSO5·KHSO4·K2SO4

Spectrum™ Chemical Oxone(R), Monopersulfate Compound , also known as potassium peroxymonosulfate or MPS, is the potassium salt of peroxymonosulfuric acid and used as an oxidizing agent. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 37222-66-5
Molecular Formula H2K4O13S3
Molecular Weight (g/mol) 462.58
InChI Key TZTINBNQGPHMBC-UHFFFAOYSA-J
IUPAC Name tetrapotassium oxidanesulfonoperoxoate hydrogen sulfate sulfate
SMILES [K+].[K+].[K+].[K+].OS([O-])(=O)=O.[O-]S([O-])(=O)=O.OS(=O)(=O)O[O-]

Specifications

CAS Max % 1
Quantity 100 g
UN Number UN3260
Grade Reagent
Packaging Poly Bottle