missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Yeast and Mold Agar (Dehydrated)

Catalog No. OXCM0920B
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0920B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXCM0920B Supplier Thermo Scientific™ Supplier No. CM0920B
Only null left

Isolate yeasts and molds within the brewing industry with this medium.

Isolate yeasts and molds with Thermo Scientific™ Oxoid™ Yeast and Mold Agar (Dehydrated). The medium is based on the formulation described by Wickerham1,2 and is formulated specifically for suitability within the brewing industry, for detection of both Saccharomyces and non-saccharomyces wild yeasts in the presence of culture yeasts.

The medium utilizes the different sensitivities of wild yeast vs. culture ale or lager yeast to the inhibitory action of copper. This enables wild yeasts to grow on the medium while suppressing the growth of culture yeast.

The medium is highly nutritious for the growth of yeasts and molds. Peptones and yeast extracts provide nitrogenous compounds and vitamin B complex. Dextrose acts as the energy source. Yeast and Mold Agar is optimized to for addition of copper usually within the range 50-300 mg/L depending on the sensitivity of the culture yeast to copper, as recommended by the Institute of Brewing Methods of analysis3.

The complete medium is recommended for examination for the microbiological quality of beers in process, pitching yeasts and packaged beers.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications

Description Yeast and Mold Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 12.2L medium

For Laboratory Use Only