missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Sulphonamide Disc

Catalog No. OXDD0011T
Change view
Click to view available options
Quantity:
50 Disks/Cartridge
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXDD0011T 50 Disks/Cartridge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXDD0011T Supplier Thermo Scientific™ Supplier No. DD0011T

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ Oxoid Sulphonamide Disc is used for identification of Gardnerella vaginalis.

Thermo Scientific™ Oxoid Sulphonamide Disc is used for identification of Gardnerella vaginalis.

  • 100μg
  • 50 Discs/Cartridge

Specifications

Content And Storage 2°C to 8°C
Description Sulphonamide Disk
Detectable Analytes G. vaginalis
Format Disk, Cartridge
Type Sulphonamide Disk
Packaging Type 1 Cart./Pk.
Quantity 50 Disks/Cartridge