Learn More
Thermo Scientific™ Oxoid™ SPS Disc
Thermo Scientific™ Oxoid SPS Disc is used for presumptive identification of Peptostreptococcus anaerobius.
Supplier:  Thermo Scientific™ DD0016T
Catalog No. OXDD0016T
Only null left
Description
Description
Thermo Scientific™ Oxoid SPS Disc is used for presumptive identification of Peptostreptococcus anaerobius.
Susceptibility disc format
Specifications
Specifications
SPS Disk
Susceptibility Disk
50 Disks/Cartridge
P. anaerobius
SPS Disk
Product Suggestions
Product Suggestions