Learn More
Thermo Scientific™ Oxoid™ Cartridge Container With Desiccant
Catalog No. OXHP0055A
Change view
Click to view available options
Quantity:
1/Pk.
Catalog No. Quantity
OXHP0055A 1/Pk.
1 options
Catalog No. OXHP0055A Supplier Thermo Scientific™ Supplier No. HP0055A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Maintain a low moisture environment while safely and conveniently storing unopened Antimicrobial Susceptibility Testing discs.
Maintain a low moisture environment while safely and conveniently storing unopened Antimicrobial Susceptibility Testing discs in the Thermo Scientific™ Oxoid™ Cartridge Container With Desiccant.
One of the simplest and most widely used methods for susceptibility testing, disc diffusion is extremely flexible, offering a wide variety of antibiotics and concentrations which can easily be changed as required.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Description Oxoid Cartridge Container With Desiccant
Product Type Cartridge Container with Desiccant
Quantity 1/Pk.