missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ ONPG Disc

Catalog No. OXDD0013T
Change view
Click to view available options
Quantity:
50 Discs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXDD0013T 50 Discs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXDD0013T Supplier Thermo Scientific™ Supplier No. DD0013T

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ Oxoid ONPG Disc is used for rapid detection of beta-galactosidase activity.

Thermo Scientific™ Oxoid ONPG Disc is used for rapid detection of beta-galactosidase activity.

Specifications

Content And Storage 2°C to 8°C
Format Disk
Type ONPG Disk
Includes ONPG Disk
Quantity 50 Discs