Learn More
Thermo Scientific™ Oxoid™ Nitrocefin Solution
Catalog No. OXSR0112C
Change view
Click to view available options
Quantity:
10 Vial Set/Pk.
Catalog No. Quantity
OXSR0112C 10 Vial Set/Pk.
1 options
Catalog No. OXSR0112C Supplier Thermo Scientific™ Supplier No. SR0112C
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Thermo Scientific™ Oxoid Nitrocefin is used for detection of beta-lactamase production.
Thermo Scientific™ Oxoid Nitrocefin is used for detection of beta-lactamase production.
  • Reconstitutes to 500μg/mL
  • Contents: 5 vials lyophilized nitrocefin (1mg) and 5 vials rehydrating fluid (1.9mL)
Specifications
Description Nitrocefin
Format Vial
Includes 5 vials lyophilized nitrocefin (1mg) and 5 vials rehydrating fluid (1.9mL)
Packaging Type 2 x 5 Vials
Quantity 10 Vial Set/Pk.