Learn More
Thermo Scientific™ Oxoid™ Moxifloxacin Antimicrobial Susceptibility discs
Catalog No. OXCT1633B
Change view
Click to view available options
Concentration:
5 μg
Catalog No. Concentration
OXCT1633B 5 μg
1 options
Catalog No. OXCT1633B Supplier Thermo Scientific™ Supplier No. CT1633B
Only null left
Determine the susceptibility of microorganisms using manual AST methods and a full range of high-quality, easy-to-use antimicrobial susceptibility testing discs.
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Moxifloxacin Antimicrobial Susceptibility discs.
Ease of use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results, but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.
Specifications
Antimicrobial Agents Moxifloxacin
Certifications/Compliance Complies with CLSI, EUCAST, SFM guidelines
Concentration 5 μg
Description Oxoid Moxifloxacin Antimicrobial Susceptibility Discs
Antibiotic Code MXF
Product Type Antimicrobial Susceptibility Disc
Test Type Antimicrobial Susceptibility
No. per Pack 5 Cartridges/Pk.
Packaging Type 50 Discs/Cartridge
Quantity 50 Discs/Cartridge