missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Microbact™ Staphylococcal 12S Kit

Catalog No. OXMB1561A
Change view
Click to view available options
Quantity:
20 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXMB1561A 20 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXMB1561A Supplier Thermo Scientific™ Supplier No. MB1561A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Simplify the identification of staphylococci, including staphylococcus aureus and coagulase-negative staphylococci (CNS), with the Thermo Scientific™ Oxoid™ Microbact™ Staphylococcal 12S Kit.

Simplify the identification of staphylococci, including staphylococcus aureus and coagulase-negative staphylococci (CNS), with the Thermo Scientific™ Oxoid™ Microbact™ Staphylococcal 12S Kit. Each strip incorporates 12 tests based on sugar utilization and colorimetric enzyme detection. The sugar utilization tests rely on a pH indicator color change, while the enzyme detection substrates produce a colored end product or react with an added indicator. Each Staphylococcus spp. produces a different pattern of reactions, allowing the definitive identification of 22 clinically relevant species.

Coagulase-negative staphylococci have become important nosocomial pathogens due, in part, to the increased use of immunosuppressive regimes and indwelling medical devices. As a result, it has become imperative for clinical microbiology laboratories to identify CNS and other staphylococci to species level8. With this kit, Staphylococcus spp. are quickly and accurately identified so that proper, fast treatment for the patient may follow.

  • Fast: Definitive identifications in less than 24 hours
  • Simple: Simple test strip format
  • Easy-to-Read and Interpret: Results are clearly visible as distinct color reactions that can be interpreted using the Microbact Identification Package (MB1244A)
  • Comprehensive: Identifies 22 clinically significant Staphylococcus, including coagulase-negative and coagulase-positive staphylococci
  • Reliable: A combination of sugar utilization and colorimetric enzyme detection substrates based upon proven, published methods1-6

Additional items required: Microbact Software Program (MB1244A), Mineral Oil (MB1093A), Fast Blue Reagent (MB1588A).

Not all products are available for sale in all territories. Please inquire.

Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications

Description Microbact Staphylococcal 12S Kit
Detectable Analytes Staphylococcus species
Type Staphylococcus aureus Test Kit
Packaging Type 20 Tests/Kit
Quantity 20 Tests/Kit
Species Staphylococcus
Target Organism Class Staphylococcus aureus