Learn More
Thermo Scientific™ Oxoid™ Microbact™ Staph Fast Blue Reagent Set
Catalog No. OXMB1588A
Change view
Click to view available options
Quantity:
2 x 10mL Bottles
Catalog No. Quantity
OXMB1588A 2 x 10mL Bottles
1 options
Catalog No. OXMB1588A Supplier Thermo Scientific™ Supplier No. MB1588A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Simplify the identification of staphylococci, including staphylococcus aureus and coagulase-negative staphylococci, with the Thermo Scientific™ Oxoid™ Microbact™ Staph Fast Blue Reagent Set.
Simplify the identification of staphylococci, including staphylococcus aureus and coagulase-negative staphylococci (CNS), with the Thermo Scientific™ Oxoid™ Microbact™ Staph Fast Blue Reagent Set. This set is used in the Beta-galactosidase test for fast, reliable and easy-to-read results.
Used in the Beta-galactosidase test, this reagent set allows for quick, reliable and easy identification of staphylococci, including staphylococcus aureus and coagulase-negative staphylococci (CNS).
Not all products are available for sale in all territories. Please inquire.
Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Description Microbact Staph Fast Blue Reagent Set
Detectable Analytes Staphylococcus species
Type Reagent Set for Staphylococcus aureus Test Kit
Packaging Type 1 Box
Quantity 2 x 10mL Bottles
Target Organism Class Staphylococcus aureus