missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Microbact™ GNB Kit

Catalog No. OXMB1132A
Change view
Click to view available options
Description:
Microbact GNB 12A Kit
Quantity:
60 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Quantity
OXMB1132A Microbact GNB 12A Kit 60 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXMB1132A Supplier Thermo Scientific™ Supplier No. MB1132A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Simplify the identification of Enterobacteriaceae and common miscellaneous Gram-negative bacilli with this biochemical identification system.

Simplify the identification of aerobic and facultatively anaerobic Gram-negative bacteria (Enterobacteriaceae and miscellaneous Gram-negative bacteria) with the Thermo Scientific™ Oxoid™ Microbact™ GNB Kit – a complete, self-contained biochemical based identification system. The kit uses 24 different biochemical tests in microplate format to produce easy-to read, distinct color reactions, generally following overnight incubation.

Gram-negative bacteria are of significant concern in both clinical infections and food contamination. Identification of the causative agent is crucial in the clinical setting to identify the cause of infection and to establish an appropriate treatment regime. In the food industry, biochemical identification can help to trace the source of contamination. With this GNB Kit, Enterobacteriaceae and common miscellaneous Gram-negative bacilli are quickly and accurately identified so that proper, fast and appropriate action may follow.

  • Fast: Most results available overnight
  • Simple and Economical: Simple microplate format
  • Easy-to-Read and Interpret: Results are clearly visible as distinct color reactions that can be interpreted using the Microbact Identification Package (MB1244A)
  • Comprehensive: Identifies >100 Gram-negative bacteria, more unusual Enterobacteriaceae and oxidase-positive, Gram-negative bacteria
  • Flexible: Use single strips or combine systems for more complex identifications. Ideal for use in clinical or food microbiology laboratories
  • Reliable: A comprehensive range of biochemical tests based on published reference methodologies1-4

Additional items required: Mineral Oil (MB1093A), Oxidase Strips (MB0266A), Microbact Software Program (MB1244A), Indole Reagent (MB0209A), VPI Reagent (MB0181A), VPII Reagent (MB0184A), TDA Reagent (MB0180A), NIT A Reagent (MB0186A), NIT B Reagent (MB0187A). The six reagents above are also available as Reagent Set D (MB1082A).

Not all products are available for sale in all territories. Please inquire.

Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications

Description Microbact GNB 12A Kit
Detectable Analytes Enterobacteriaceae
Packaging Type 1 Box
Quantity 60 Tests/Kit