missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Metronidazole Disc

Catalog No. OXDD0008T
Change view
Click to view available options
Quantity:
50 Disks
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXDD0008T 50 Disks
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXDD0008T Supplier Thermo Scientific™ Supplier No. DD0008T

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ Oxoid Metronidazole Disc is used for presumptive identification of Gardnerella vaginalis.

Thermo Scientific™ Oxoid Metronidazole Disc is used for presumptive identification of Gardnerella vaginalis.

Specifications

Content And Storage -20°C
Description Metronidazole Disk
Detectable Analytes G. vaginalis
Format Disk, Cartridge
Type Metronidazole Disk
Packaging Type 1 Cart./Pk.
Quantity 50 Disks