Learn More
Thermo Scientific™ Membrane Lauryl Sulphate Broth (Dehydrated)
Description
Enumerate coliform organisms and Escherichia coli in water by membrane filtration with this medium.
Enumerate coliform organisms and Escherichia coli in water by membrane filtration with Thermo Scientific™ Oxoid™ Membrane Lauryl Sulphate Broth (Dehydrated).
Membrane Lauryl Sulphate Broth has been recommended1,2 as a standard medium for the enumeration of coliform organisms and Escherichia coli from water and sewage by membrane filtration technique.
Membrane Lauryl Sulphate Broth is similar to Membrane Enriched Teepol Broth except teepol has been replaced by sodium lauryl sulphate. Sodium lauryl sulphate was found to be an adequate reproducible substitute1.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Description | Membrane Lauryl Sulphate Broth |
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| Quantity | 500 g |
| Yield | For 6.6L medium |
For Laboratory Use Only