Learn More
Thermo Scientific™ Oxoid™ Malt Extract Agar (Dehydrated)
Detect, isolate and enumerate yeasts and molds with typical morphology and pigmentation using medium.
Supplier:  Thermo Scientific™ CM0059B
 View more versions of this product
Catalog No. OXCM0059B
Only null left
Explore available promotions
Description
Description
Detect, isolate and enumerate yeasts and molds with Thermo Scientific™ Oxoid™ Malt Extract Agar (Dehydrated). Mycological peptone in the medium gives a luxuriant growth of yeasts and molds with typical morphology and pigmentation.
Malt Extract Agar is similar to the formula of Galloway and Burgess1 used for the detection, isolation, and enumeration of yeasts and molds. For mycological counts, it may be desirable to prepare the more acid medium in order to suppress bacterial growth.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
Specifications
Malt Extract Agar
Agar
For 10.0L medium
Powder
500 g
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Laboratory Use Only