Learn More
Thermo Scientific™ Oxoid™ Gentamicin Antimicrobial Susceptibility Discs
Determine the susceptibility of microorganisms using manual AST methods and a full range of high-quality, easy-to-use antimicrobial susceptibility testing discs.
Supplier:  Thermo Scientific™ CT0794B
 View more versions of this product
Catalog No. OXCT0794B
Only null left
Description
Description
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Gentamycin Antimicrobial Susceptibility discs.
  • Easy-to-read labeling
  • Shatterproof, spring-loaded cartridges
  • End-of-cartridge indicator
  • Reproducible performance
Ordering Alerts:
50 Discs/Cartridge; 5 Cartridges/Package. Sold by 5 Cartridges/Package only.
Specifications
Chemical Identifiers
Gentamicin
CN
Antimicrobial Susceptibility
50 Discs/Cartridge
Oxoid Gentamicin Antimicrobial Susceptibility Discs
Antimicrobial Susceptibility Discs
5 Cartridges/Pk.
50 Discs/Cartridge
Specifications
Gentamicin
120 μg
CN
Antimicrobial Susceptibility
50 Discs/Cartridge
Complies with CLSI guidelines
Oxoid Gentamicin Antimicrobial Susceptibility Discs
Antimicrobial Susceptibility Discs
5 Cartridges/Pk.
50 Discs/Cartridge
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Accessories