Learn More
Thermo Scientific™ Oxoid™ Gentamicin Antimicrobial Susceptibility Discs
Catalog No. OXCT0024B
Change view
Click to view available options
Concentration:
10 μg
120 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
OXCT0024B 10 μg
OXCT0794B 120 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. OXCT0024B Supplier Thermo Scientific™ Supplier No. CT0024B
Only null left
Determine the susceptibility of microorganisms using manual AST methods and a full range of high-quality, easy-to-use antimicrobial susceptibility testing discs.
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Gentamycin Antimicrobial Susceptibility discs.
  • Easy-to-read labeling
  • Shatterproof, spring-loaded cartridges
  • End-of-cartridge indicator
  • Reproducible performance
Ordering Alerts:
50 Discs/Cartridge; 5 Cartridges/Package. Sold by 5 Cartridges/Package only.
Specifications
Antimicrobial Agents Gentamicin
Certifications/Compliance Complies with CLSI, DIN, BSAC, EUCAST guidelines
Concentration 10 μg
Description Oxoid Gentamicin Antimicrobial Susceptibility Discs
Antibiotic Code CN
Product Type Antimicrobial Susceptibility Discs
Test Type Antimicrobial Susceptibility
No. per Pack 5 Cartridges/Pk.
Packaging Type 50 Discs/Cartridge
Quantity 50 Discs/Cartridge