Learn More
Thermo Scientific™ Gamma-Irradiated Tryptone Soya Broth (Dehydrated)
Cultivate a wide range of organisms with this gamma-irradiated medium that is particularly suitable for the Media Fill Tests.
Supplier: Thermo Scientific™ CM1016T
Description
Cultivate a wide range of organisms with nutritious Thermo Scientific™ Oxoid™ Gamma-Irradiated Tryptone Soya Broth (TSB) (Dehydrated) which has been given a sterilizing dose of irradiation. Gamma-Irradiated Tryptone Soya Broth is particularly suitable for the microbiological validation of aseptic filling (Media Fill Test).
Media Fill Test is a validation method used to assess the performance of aseptic processing in the pharmaceutical industry. Pharmaceutical products are substituted by Gamma irradiated TSB and the medium is subjected to exactly the same conditions as the products, including filling and closing, to ensure that there is no microbial contamination occurring during the process.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
| Tryptone Soya Broth, Gamma-Irradiated | |
| Powder | |
| Dehydrated Culture Media | |
| 5 kg |
For Laboratory Use Only