missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Gamma-Irradiated Tryptone Soya Broth (Dehydrated)

Catalog No. OXCM1016B
Change view
Click to view available options
Quantity:
500 g
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM1016B 500 g
OXCM1016T 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. OXCM1016B Supplier Thermo Scientific™ Supplier No. CM1016B
Only null left

Cultivate a wide range of organisms with this gamma-irradiated medium that is particularly suitable for the Media Fill Tests.

Cultivate a wide range of organisms with nutritious Thermo Scientific™ Oxoid™ Gamma-Irradiated Tryptone Soya Broth (TSB) (Dehydrated) which has been given a sterilizing dose of irradiation. Gamma-Irradiated Tryptone Soya Broth is particularly suitable for the microbiological validation of aseptic filling (Media Fill Test).

Media Fill Test is a validation method used to assess the performance of aseptic processing in the pharmaceutical industry. Pharmaceutical products are substituted by Gamma irradiated TSB and the medium is subjected to exactly the same conditions as the products, including filling and closing, to ensure that there is no microbial contamination occurring during the process.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

Specifications

Description Tryptone Soya Broth, Gamma-Irradiated
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g

For Laboratory Use Only