Learn More
Thermo Scientific™ Oxoid™ Demi-Fraser Broth (without Ferric Ammonium Citrate) (Dehydrated)
Isolate Listeria spp. from food and environmental samples with this medium.
$389.51 - $1336.33
Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
OXCM1053B
View Documents
Thermo Scientific™
CM1053B
500 g
Each for $389.51
Only null left
OXCM1053R
View Documents
Thermo Scientific™
CM1053R
2.5 kg
Each for $1,336.33
Only null left
Description
Description
Isolate Listeria spp. from food and environmental samples with Thermo Scientific™ Demi-Fraser Broth (without Ferric Ammonium Citrate) (Dehydrated), when used as the secondary enrichment medium in methodology for Listeria isolation. This medium is a modification of Fraser Broth and contains half of the concentration of nalidixic acid and acriflavin hydrochloride to aid in the recovery of stressed cells. The removal of ferric ammonium citrate from the formula aids in subculture from broth to imaging instrumentation or Polymerase Chain Reaction-based assay machines.
Listeria monocytogenes is the most common pathogenic Listeria spp. and has been shown to be pathogenic to both humans and animals. Use Demi-Fraser Broth (without Ferric Ammonium Citrate) for isolation of Listeria species from food and environmental samples when used as the secondary enrichment medium in methodology for Listeria isolation.
  • Helps in the recovery of stressed cells
  • Allows subculture from the broth to imaging instrumentation or PCR based assay machines
Not all products are available for sale in all territories. Please inquire.
Remel™ and Thermo Scientific™ products are now part of the Thermo Scientific brand
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
For Laboratory Use Only