Learn More
Thermo Scientific™ Oxoid™ CampyGen™ Compact Sachet
Catalog No. OXCN0020C
Change view
Click to view available options
Quantity:
20/Pk.
Catalog No. Quantity
OXCN0020C 20/Pk.
1 options
Catalog No. OXCN0020C Supplier Thermo Scientific™ Supplier No. CN0020C
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Produce suitable gaseous atmosphere for growth of microaerophilic microorganisms with the Oxoid CampyGen Compact Sachet.
Produce suitable gaseous atmosphere for growth of microaerophilic microorganisms with the Oxoid CampyGen Compact Sachet.
  • For use in Compact Plastic Pouch
  • Each pouch holds 2 standard size petri dishes
  • No water or catalyst required
  • No hydrogen gas produced
  • Use with Compact Plastic Pouch Part No. AG0020C and AnaeroGen™ Compact Sealing Clip, Part No. AN0005C
Specifications
Type CampyGen Compact Sachet
Format Compact Gas Generation Sachet
Product Line Thermo Scientific Oxoid
Quantity 20/Pk.