Learn More
Thermo Scientific™ Brain Heart Infusion Broth (Dehydrated)
Cultivate streptococci, Neisseria and other fastidious organisms with this highly nutritious medium.
Supplier: Thermo Scientific™ CM1135B
Description
Cultivate streptococci, Neisseria and other fastidious organisms with Thermo Scientific™ Oxoid™ Brain Infusion Broth (Dehydrated). The medium is highly nutritious and well buffered to support the growth of wide variety of organisms. It is recommended for blood culture work for the isolation and cultivation of pathogenic fungi.
Cultivate a wide variety of organisms with Brain Infusion Broth:
- Use as a growth and suspension medium for staphylococci when testing coagulase production.
- Add 10% defibrillated blood for isolation and cultivation of pathogenic fungi.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
| Brain Heart Infusion Broth | |
| Dehydrated Culture Media | |
| For 13.5L medium |
| Powder | |
| 500 g |