Learn More
Thermo Scientific™ Brain Heart Infusion Agar (Dehydrated)
Cultivate fastidious organisms with this highly nutritious medium.
Supplier: Thermo Scientific™ CM1136B
Description
Cultivate fastidious organisms with highly nutritious Thermo Scientific™ Oxoid™ Brain Heart Infusion Agar (Dehydrated). Brain Heart Infusion Agar is recommended for the cultivation of streptococci, Neisseria and other fastidious organisms.
Cultivate a wide variety of organisms with Brain Heart Infusion Agar.
- Recommended for the isolation if Neisseria gonorrhoeae1.
- With addition of blood and antibiotics the medium is suitable for the isolation of the tissue phase of Histoplasma capsulatum and other pathogenic fungi including Coccidiodes immitis2, 3.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
| Industrial Reference: AFNOR, BSI, FDA, IDF, ISO, USDA. | |
| Powder | |
| 500 g |
| Brain Heart Infusion Agar | |
| Agar | |
| For 10.6L medium |