missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Baird-Parker RPF Agar Base
Selective medium for the isolation and enumeration of coagulase positive staphylococci from food.
Supplier: Thermo Scientific™ CM0961B
Description
The Oxoid Baird-Parker RPF Agar Base is a selective medium for the isolation and enumeration of coagulase positive staphylococci from food.
- For use with RPF supplement, Part No. SR0122A
- Do not use Egg Yolk Tellurite Emulsion
Specifications
| Industrial Reference: ISO, NMKL. | |
| Powder | |
| 500 g |
| Baird-Parker Agar Base, RPF | |
| Agar | |
| For 7.1L medium |
For Laboratory Use Only