Learn More
Thermo Scientific™ Oxoid™ Antimicrobial Susceptibility disc Dispenser
Accurately place antimicrobial susceptibility discs onto your media of choice when carrying out manual Antimicrobial Susceptibility Testing methods.
Specifications
| Certifications/Compliance | For use only with Oxoid Antimicrobial Susceptibility Discs |
|---|---|
| Description | Oxoid Antimicrobial Susceptibility Disc Dispenser |
| Product Type | Antimicrobial Susceptibility Disc Dispenser |
| Packaging Type | 1 Dispenser/Pk. |
| Catalog Number | Mfr. No. | No. of Places | For Use With (Equipment) | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | No. of Places | For Use With (Equipment) | Price | Quantity & Availability | ||||
|
OXST8100
|
Thermo Scientific™
ST8100 |
8 Oxoid Disc Cartridges | 100mm diameter plates |
N/A
|
|
||||
Description
Accurately place antimicrobial susceptibility discs onto your media of choice when carrying out manual Antimicrobial Susceptibility Testing methods using a self-tamping Thermo Scientific™ Oxoid™ Antimicrobial Susceptibility disc Dispenser.
One of the simplest and most widely used methods for susceptibility testing, disc diffusion is extremely flexible, offering a wide variety of antibiotics and concentrations which can easily be changed as required.disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
- Inoculum level
- Presence of contamination
- Resistant mutants
- Beta-lactamase activity
- Antagonism and synergy between antibiotics
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
| For use only with Oxoid Antimicrobial Susceptibility Discs | |
| Oxoid Antimicrobial Susceptibility Disc Dispenser | |
| Antimicrobial Susceptibility Disc Dispenser | |
| 1 Dispenser/Pk. |
For use only with Oxoid™ Antimicrobial Susceptibility discs.