Learn More
Thermo Scientific™ Remel™ Antimicrobial Susceptibility disc Dispenser, 150 mm, 12-place
Description
Accurately place antimicrobial susceptibility discs onto your media of choice when carrying out manual Antimicrobial Susceptibility Testing methods using a self-tamping Thermo Scientific™ Remel™ Antimicrobial Susceptibility disc Dispenser.
One of the simplest and most widely used methods for susceptibility testing, disc diffusion is extremely flexible, offering a wide variety of antibiotics and concentrations which can easily be changed as required.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
- Inoculum level
- Presence of contamination
- Resistant mutants
- Beta-lactamase activity
- Antagonism and synergy between antibiotics
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Certifications/Compliance | For use only with Oxoid Antimicrobial Susceptibility Discs |
| Description | Remel Antimicrobial Susceptibility Disc Dispenser |
| Product Type | Antimicrobial Susceptibility Disc Dispenser |
| No. of Places | 12-place |
| Quantity | 1 Dispenser/Pk. |