Learn More
Thermo Scientific™ Oxoid™ AnaeroJar™ Base, Jar
Catalog No. OXAG0026A
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
OXAG0026A 1 Ea.
1 options
Catalog No. OXAG0026A Supplier Thermo Scientific™ Supplier No. AG0026A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Oxoid AnaeroJar Base is a polycarbonate jar base for use with AnaeroJar 2.5L, Part No. AG0025A.
Oxoid AnaeroJar Base is a polycarbonate jar base for use with AnaeroJar 2.5L, Part No. AG0025A.
Specifications
Type AnaeroJar Base
Format Oval Jar
Product Line Thermo Scientific Oxoid
Quantity 1 Ea.