Learn More
Thermo Scientific™ Oxoid™ Anaerobic System Schrader Valve Chuck plus Clip, Pressure Gauge
Catalog No. OXHP0020A
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
OXHP0020A 1 Ea.
1 options
Catalog No. OXHP0020A Supplier Thermo Scientific™ Supplier No. HP0020A
Only null left
Oxoid Anaerobic System Schrader Valve Chuck plus Clip is for use with Thermo Scientific™ Oxoid 3.5L Anaerobic Jar, Part No. HP0011A.
Oxoid Anaerobic System Schrader Valve Chuck plus Clip is for use with Thermo Scientific™ Oxoid 3.5L Anaerobic Jar, Part No. HP0011A.
Specifications
Type Anaerobic System Schrader Valve Chuck plus Clip
Includes Pressure Gauge
Product Line Thermo Scientific Oxoid
Quantity 1 Ea.