Learn More
Thermo Scientific™ Oxoid An-Ident Discs
Presumptive identification for Gram-negative anaerobic organisms
Supplier:  Thermo Scientific™ DD0006A
Catalog No. OXDD0006A
Only null left
Includes:
  • Erythromycin, 60μg
  • Rifampin, 15μg
  • Colistin, 10μg
  • Penicillin, 2U
  • Kanamycin, 1000μg
  • Vancomycin, 5μg
Description
Description
Order Info
Supplied 50 disks per cartridge; 6 cartridges/set.
Specifications
Specifications
Gram-negative anaerobic organisms
Oxoid An-Ident Discs
Erythromycin 60μg, Rifampin 15μg, Colistin 10μg, Penicillin 2U, Kanamycin 1000μg, Vancomycin 5μg
50 Disks
Product Suggestions
Product Suggestions