missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Alternative Thioglycollate Broth

Catalog No. oxcm0391b
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0391B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXCM0391B Supplier Thermo Scientific™ Supplier No. CM0391B
Only null left

Oxoid Thioglycollate Broth, Alternative is a non-viscous thioglycollate medium is used for the culture of turbid biological products.

Oxoid Thioglycollate Broth, Alternative is a non-viscous thioglycollate medium is used for the culture of turbid biological products.

Specifications

Certifications/Compliance Industrial Reference: USP Alternative.
Description Thioglycollate Broth (USP alternative)
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 17.2L medium