missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Agar Technical

Catalog No. p-6078071
Change view
Click to view available options
Quantity:
5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXLP0013T 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXLP0013T Supplier Thermo Scientific™ Supplier No. LP0013T
Only null left

The Oxoid Agar Technical (Agar No. 3) is a general-purpose bacteriological-grade agar.

Oxoid Agar Technical (Agar No. 3) is a general-purpose bacteriological-grade agar.

  • Normal working concentration 1.2%

Specifications

Description Agar Tech (agar No 3) 5 Kg
Form Powder
Product Type Agar
Quantity 5 kg

For Laboratory Use Only