missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Agar Bacteriological

Catalog No. oxlp0011t
Change view
Click to view available options
Quantity:
500 g
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXLP0011T 5 kg
OXLP0011B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. OXLP0011T Supplier Thermo Scientific™ Supplier No. LP0011T
Only null left

Oxoid Agar Bacteriological (Agar No. 1) is a clear purified high gel strength bacteriological agar compatible with all culture media.

Oxoid Agar Bacteriological (Agar No. 1) is a clear purified high gel strength bacteriological agar compatible with all culture media.

Specifications

Description Agar No. 1, bacteriological
Form Powder
Product Type Agar
Quantity 5 kg