missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxo[5,10,15,20-tetra(4-pyridyl)porphyrinato]titanium(IV) 90.0+%, TCI America™
Specifications
Specifications
| Color | Purple-Red |
| Quantity | 100 mg |
| Formula Weight | 680.55 |
| Percent Purity | ≥90.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Oxo[5,10,15,20-tetra(4-pyridyl)porphyrinato]titanium(IV) [Determination of Glucose in Serum and Urin |
Safety and Handling
| TSCA | No |