missing translation for 'onlineSavingsMsg'
Learn More

Oxiracetam 96.0+%, TCI America™

Catalog No. p-7134316
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03981G 1 g
O03985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O03981G Supplier TCI America Supplier No. O03981G
Only null left

Chemical Identifiers

CAS 62613-82-5
Molecular Formula C6H10N2O3
Molecular Weight (g/mol) 158.157
MDL Number MFCD00242951
InChI Key IHLAQQPQKRMGSS-UHFFFAOYSA-N
Synonym 2-(4-Hydroxy-2-oxo-1-pyrrolidinyl)acetamide
PubChem CID 4626
IUPAC Name 2-(4-hydroxy-2-oxopyrrolidin-1-yl)acetamide
SMILES C1C(CN(C1=O)CC(=O)N)O

Specifications

Melting Point 172°C
Color White
Quantity 1 g
Formula Weight 158.16
Percent Purity ≥96.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material Oxiracetam
RTECSNumber UX9656638
TSCA No
Recommended Storage Refrigerator