missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxiracetam 96.0+%, TCI America™
Specifications
Specifications
| Melting Point | 172°C |
| Color | White |
| Quantity | 1 g |
| Formula Weight | 158.16 |
| Percent Purity | ≥96.0% (GC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Oxiracetam |
Safety and Handling
| RTECSNumber | UX9656638 |
| TSCA | No |
| Recommended Storage | Refrigerator |